(4-Chloro-3-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

68756-500MG 68756-1G 68756-5G 68756-25G
77333-298EA 220.42 USD
77333-298 77357-484 77369-318 77390-356
(4-Chloro-3-fluorophenyl)trimethylsilane ≥97%
(4-Chloro-3-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₂ClFSi
MW: 202.728 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201834
CAS Number: 845823-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR