(4-Bromo-3-chloro-2-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

68754-500MG 68754-1G 68754-5G 68754-25G
77333-294EA 134.93 USD
77333-294 77344-486 77369-314 77390-352
(4-Bromo-3-chloro-2-fluorophenyl)trimethylsilane ≥97%
(4-Bromo-3-chloro-2-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₁BrClFSi
MW: 281.625 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763601
CAS Number: 1864257-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR