(3-Chloro-5-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

68750-500MG 68750-1G 68750-5G 68750-25G
77333-292EA 220.42 USD
77333-292 77357-482 77369-310 77390-350
(3-Chloro-5-fluorophenyl)trimethylsilane ≥95%
(3-Chloro-5-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₂ClFSi
Storage Temperature: Ambient
MDL Number: MFCD06201838
CAS Number: 845823-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR