5-(2-Chloro-6-methoxyphenyl)thiazol-2-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
68729-500MG
68729-1G
68729-5G
68729-25G
77333-288EA
184.37
USD
77333-288
77347-320
77369-306
77390-348
5-(2-Chloro-6-methoxyphenyl)thiazol-2-amine ≥97%
5-(2-Chloro-6-methoxyphenyl)thiazol-2-amine
Precaución: for Research Use Only
Formula:
C₁₀H₉ClN₂OS MW: 240.709 g/mol Storage Temperature: Ambient |
CAS Number:
1600891-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...