1-Fluoro-3-iodo-2-isopropoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

68704-500MG 68704-1G 68704-5G 68704-25G
77333-278EA 116.39 USD
77333-278 77343-456 77369-300 77381-410
1-Fluoro-3-iodo-2-isopropoxy-5-methylbenzene ≥97%
1-Fluoro-3-iodo-2-isopropoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO
MW: 294.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763574
CAS Number: 2807471-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR