1-(4-Chloro-3-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

68611-500MG 68611-1G 68611-5G 68611-25G
77333-268EA 103 USD
77333-268 77343-050 77369-290 77381-404
1-(4-Chloro-3-methoxyphenyl)ethanol ≥95%
1-(4-Chloro-3-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD23752186
CAS Number: 1529744-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR