1-(3-Bromo-2-fluoro-5-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

68601-500MG 68601-1G 68601-5G 68601-25G
77333-266EA 202.91 USD
77333-266 77357-464 77369-288 77390-336
1-(3-Bromo-2-fluoro-5-methylphenyl)ethanol ≥97%
1-(3-Bromo-2-fluoro-5-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD28130743
CAS Number: 1780677-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR