4'-Phenoxy-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,2,2-Trifluoro-1-(4-phenoxyphenyl)ethanone

68580-1G 68580-5G 68580-25G
77342-130EA 114.33 USD
77342-130 77369-286 77381-394
4'-Phenoxy-2,2,2-trifluoroacetophenone ≥95%
4'-Phenoxy-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O₂
MW: 266.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042392
CAS Number: 70783-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR