1-Bromo-2,3-dimethyl-4-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

68570-1G 68570-5G 68570-25G
77340-814EA 60.77 USD
77340-814 77369-282 77381-390
1-Bromo-2,3-dimethyl-4-propoxybenzene ≥95%
1-Bromo-2,3-dimethyl-4-propoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763493
CAS Number: 2807448-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR