(((3-Fluoro-1,2-phenylene)bis(oxy))bis(methylene))dibenzene ≥97%

Supplier: AOBChem USA

Warning

68565-500MG 68565-1G 68565-5G 68565-25G
77333-262EA 128.75 USD
77333-262 77344-286 77369-280 77381-388
(((3-Fluoro-1,2-phenylene)bis(oxy))bis(methylene))dibenzene ≥97%
(((3-Fluoro-1,2-phenylene)bis(oxy))bis(methylene))dibenzene

Precaución: for Research Use Only

Formula: C₂₀H₁₇FO₂
MW: 308.346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763491
CAS Number: 103138-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR