3-(Benzyloxy)-4-ethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

68550-500MG 68550-1G 68550-5G 68550-25G
77333-258EA 228.66 USD
77333-258 77357-454 77369-276 77390-332
3-(Benzyloxy)-4-ethoxy-2-fluorobenzoic acid ≥97%
3-(Benzyloxy)-4-ethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅FO₄
Storage Temperature: Ambient
MDL Number: MFCD34763476
CAS Number: 2901102-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR