2-(Benzyloxy)-1-ethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

68547-500MG 68547-1G 68547-5G 68547-25G
77333-254EA 161.71 USD
77333-254 77345-970 77369-272 77390-328
2-(Benzyloxy)-1-ethoxy-3-fluorobenzene ≥97%
2-(Benzyloxy)-1-ethoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂
MW: 246.277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763473
CAS Number: 2807440-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR