(5-Chloro-6-fluoro-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

68546-500MG 68546-1G 68546-25G 68546-5G
77333-252EA 302.82 USD
77333-252 77357-452 77390-326 77381-384
(5-Chloro-6-fluoro-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%
(5-Chloro-6-fluoro-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₉ClF₄OS
MW: 336.732 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763472
CAS Number: 2901081-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR