2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

68519-500MG 68519-1G 68519-5G
77339-716EA 616.97 USD
77339-716 77357-446 77381-378
2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
MW: 272.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763446
CAS Number: 2901099-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR