2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

68517-500MG 68517-1G 68517-5G
77339-228EA 568.56 USD
77339-228 77357-444 77381-376
2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%
2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₅
MW: 288.295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18321413
CAS Number: 1261991-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR