(2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

68515-500MG 68515-1G 68515-5G
77339-810EA 649.93 USD
77339-810 77357-442 77381-374
(2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-yl)boronic acid ≥97%
(2,3',5'-Trimethoxy-[1,1'-biphenyl]-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₇BO₅
MW: 288.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763443
CAS Number: 955085-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR