1-(2-Chloro-4-fluoro-3-(methylthio)phenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
68499-500MG
68499-1G
68499-5G
68499-25G
77333-248EA
184.37
USD
77333-248
77347-318
77369-270
77390-324
1-(2-Chloro-4-fluoro-3-(methylthio)phenyl)ethanone ≥97%
1-(2-Chloro-4-fluoro-3-(methylthio)phenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₈ClFOS MW: 218.676 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34763430 CAS Number: 2918828-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...