5-Bromo-1,2-dimethoxy-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

68469-500MG 68469-1G 68469-5G
77337-468EA 414.06 USD
77337-468 77357-432 77381-364
5-Bromo-1,2-dimethoxy-3-(methoxymethyl)benzene ≥97%
5-Bromo-1,2-dimethoxy-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₃
MW: 261.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763408
CAS Number: 2807454-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR