2-Chloro-5-ethyl-3-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

68467-500MG 68467-1G
77340-428EA 1044.42 USD
77340-428 77357-430
2-Chloro-5-ethyl-3-(methylthio)benzaldehyde ≥95%
2-Chloro-5-ethyl-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClOS
MW: 214.712 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763406
CAS Number: 2807448-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR