2-Chloro-5-ethyl-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

68465-500MG 68465-1G 68465-5G
77338-116EA 465.56 USD
77338-116 77357-428 77381-362
2-Chloro-5-ethyl-3-(methylthio)benzoic acid ≥97%
2-Chloro-5-ethyl-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂S
MW: 230.711 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763404
CAS Number: 2807448-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR