3-Chloro-2-ethoxy-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68421-500MG 68421-1G 68421-5G 68421-25G
77333-238EA 252.35 USD
77333-238 77357-420 77369-266 77390-316
3-Chloro-2-ethoxy-4-methoxybenzaldehyde ≥97%
3-Chloro-2-ethoxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763364
CAS Number: 2384684-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR