4-Chloro-2-iodo-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

68412-1G 68412-5G 68412-25G
77340-812EA 60.77 USD
77340-812 77369-264 77381-352
4-Chloro-2-iodo-1-(trifluoromethoxy)benzene ≥97%
4-Chloro-2-iodo-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃ClF₃IO
Storage Temperature: Ambient
MDL Number: MFCD20492510
CAS Number: 1263057-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR