5-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

68385-1G 68385-5G 68385-25G
77340-964EA 64.89 USD
77340-964 77369-262 77381-350
5-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid ≥97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043332
CAS Number: 1881295-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR