1-(3,5-Difluoro-4-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

68355-500MG 68355-1G 68355-5G 68355-25G
77333-234EA 120.51 USD
77333-234 77343-802 77369-260 77381-348
1-(3,5-Difluoro-4-methylphenyl)-2,2-difluoroethanone ≥95%
1-(3,5-Difluoro-4-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆F₄O
MW: 206.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763341
CAS Number: 2807439-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR