2,2-Difluoro-1-(3-fluoro-5-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

68354-500MG 68354-1G 68354-5G
77336-938EA 400.67 USD
77336-938 77357-416 77381-346
2,2-Difluoro-1-(3-fluoro-5-(trifluoromethyl)phenyl)ethanone ≥95%
2,2-Difluoro-1-(3-fluoro-5-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄F₆O
MW: 242.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28104292
CAS Number: 1823813-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR