1-(3-Bromo-2,6-difluoro-5-methoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

68351-500MG 68351-1G 68351-5G
77336-654EA 390.37 USD
77336-654 77357-412 77381-342
1-(3-Bromo-2,6-difluoro-5-methoxyphenyl)-2,2-difluoroethanone ≥95%
1-(3-Bromo-2,6-difluoro-5-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅BrF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34763340
CAS Number: 2807467-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR