3-Bromo-2-ethoxy-5-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68350-500MG 68350-1G 68350-5G
77333-232EA 332.69 USD
77333-232 77357-410 77381-340
3-Bromo-2-ethoxy-5-fluoro-1,1'-biphenyl ≥97%
3-Bromo-2-ethoxy-5-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763339
CAS Number: 2901102-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR