2-Ethoxy-5-fluoro-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

68346-500MG 68346-1G 68346-5G 68346-25G
77333-230EA 167.89 USD
77333-230 77346-228 77369-258 77390-312
2-Ethoxy-5-fluoro-3-(methylthio)benzoic acid ≥97%
2-Ethoxy-5-fluoro-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃S
Storage Temperature: Ambient
MDL Number: MFCD34763336
CAS Number: 2901098-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR