3-Bromo-2-ethoxy-5-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

68334-1G 68334-5G 68334-25G
77341-702EA 100.94 USD
77341-702 77369-250 77381-334
3-Bromo-2-ethoxy-5-fluorophenol ≥97%
3-Bromo-2-ethoxy-5-fluorophenol

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12910495
CAS Number: 1026796-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR