1-Fluoro-2-(methylsulfonyl)-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

68332-1G 68332-5G 68332-25G
77341-836EA 105.06 USD
77341-836 77369-248 77381-330
1-Fluoro-2-(methylsulfonyl)-4-(trifluoromethyl)benzene ≥97%
1-Fluoro-2-(methylsulfonyl)-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆F₄O₂S
MW: 242.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527245
CAS Number: 1824071-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR