4-Bromo-2-ethyl-3',6-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68323-500MG 68323-1G 68323-5G 68323-25G
77333-224EA 279.13 USD
77333-224 77357-404 77369-244 77390-306
4-Bromo-2-ethyl-3',6-dimethyl-1,1'-biphenyl ≥95%
4-Bromo-2-ethyl-3',6-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763324
CAS Number: 2901102-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR