4-Bromo-2-ethyl-4',6-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68320-500MG 68320-1G 68320-5G
77337-540EA 418.18 USD
77337-540 77357-402 77381-326
4-Bromo-2-ethyl-4',6-dimethyl-1,1'-biphenyl ≥95%
4-Bromo-2-ethyl-4',6-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763321
CAS Number: 2901081-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR