4-Bromo-2-ethyl-6-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68317-500MG 68317-1G 68317-5G
77335-516EA 356.38 USD
77335-516 77357-400 77381-324
4-Bromo-2-ethyl-6-methyl-1,1'-biphenyl ≥97%
4-Bromo-2-ethyl-6-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
MW: 275.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763318
CAS Number: 2901109-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR