5-Bromo-1-ethyl-2-(methoxymethyl)-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

68310-500MG 68310-1G 68310-5G
77335-722EA 363.59 USD
77335-722 77357-396 77381-320
5-Bromo-1-ethyl-2-(methoxymethyl)-3-methylbenzene ≥97%
5-Bromo-1-ethyl-2-(methoxymethyl)-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763311
CAS Number: 2807440-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR