(2,5-Dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

68309-500MG 68309-1G 68309-5G 68309-25G
77333-222EA 239.99 USD
77333-222 77357-394 77369-242 77390-304
(2,5-Dimethoxyphenyl)(methyl)sulfane ≥97%
(2,5-Dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00010653
CAS Number: 2570-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR