2-Ethoxy-3-fluoro-4-propoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

68302-500MG 68302-1G 68302-5G
77336-552EA 388.31 USD
77336-552 77357-392 77381-318
2-Ethoxy-3-fluoro-4-propoxybenzoic acid ≥95%
2-Ethoxy-3-fluoro-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₄
Storage Temperature: Ambient
MDL Number: MFCD34763305
CAS Number: 2901096-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR