5-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68298-500MG 68298-1G 68298-5G 68298-25G
77333-220EA 278.1 USD
77333-220 77357-388 77369-240 77390-302
5-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzaldehyde ≥97%
5-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₄O₂
MW: 250.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763301
CAS Number: 2901109-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR