5-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

68296-500MG 68296-1G 68296-5G
77335-660EA 362.56 USD
77335-660 77357-386 77381-314
5-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzoic acid ≥95%
5-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₄O₃
MW: 266.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763299
CAS Number: 2901098-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR