4-Bromo-2-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68287-500MG 68287-1G 68287-5G
77338-910EA 528.39 USD
77338-910 77357-384 77381-312
4-Bromo-2-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥95%
4-Bromo-2-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₉BrF₄O
MW: 349.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763290
CAS Number: 2901102-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR