4'-Bromo-2'-fluoro-3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

68285-500MG 68285-1G 68285-5G 68285-25G
77333-218EA 239.99 USD
77333-218 77357-382 77369-238 77390-300
4'-Bromo-2'-fluoro-3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%
4'-Bromo-2'-fluoro-3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrFO₂
MW: 309.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763288
CAS Number: 2901099-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR