1-Bromo-3-fluoro-2-methoxy-4-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

68268-500MG 68268-1G 68268-5G 68268-25G
77333-212EA 95.79 USD
77333-212 77342-590 77369-232 77381-310
1-Bromo-3-fluoro-2-methoxy-4-(methylsulfonyl)benzene ≥97%
1-Bromo-3-fluoro-2-methoxy-4-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₃S
Storage Temperature: Ambient
MDL Number: MFCD34763281
CAS Number: 2807445-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR