2-Fluoro-5-methyl-3-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

68240-500MG 68240-1G 68240-25G 68240-5G
77333-206EA 303.85 USD
77333-206 77357-378 77390-290 77381-308
2-Fluoro-5-methyl-3-(methylthio)phenol ≥97%
2-Fluoro-5-methyl-3-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉FOS
Storage Temperature: Ambient
MDL Number: MFCD34763261
CAS Number: 2706345-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR