1-Bromo-5-fluoro-2-methoxy-3-(methylsulfonyl)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
68227-500MG
68227-1G
68227-5G
68227-25G
77333-196EA
141.11
USD
77333-196
77344-892
77369-220
77390-282
1-Bromo-5-fluoro-2-methoxy-3-(methylsulfonyl)benzene ≥97%
1-Bromo-5-fluoro-2-methoxy-3-(methylsulfonyl)benzene
Precaución: for Research Use Only
Formula:
C₈H₈BrFO₃S MW: 283.115 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34763254 CAS Number: 2901102-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...