5-Fluoro-2-methoxy-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68223-500MG 68223-1G 68223-5G
77333-192EA 340.93 USD
77333-192 77357-374 77381-304
5-Fluoro-2-methoxy-3-(methylthio)benzaldehyde ≥97%
5-Fluoro-2-methoxy-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763250
CAS Number: 1895585-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR