5-Fluoro-2-methoxy-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

68221-500MG 68221-1G 68221-5G 68221-25G
77333-190EA 264.71 USD
77333-190 77357-372 77369-216 77390-278
5-Fluoro-2-methoxy-3-(methylthio)benzoic acid ≥97%
5-Fluoro-2-methoxy-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃S
Storage Temperature: Ambient
MDL Number: MFCD34763248
CAS Number: 1895435-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR