3-Chloro-2,6-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

68215-500MG 68215-1G 68215-5G 68215-25G
77333-184EA 120.51 USD
77333-184 77343-800 77369-212 77381-298
3-Chloro-2,6-dimethylbenzoic acid ≥95%
3-Chloro-2,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486272
CAS Number: 90649-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR