(2,6-Difluoro-3-(methoxymethyl)phenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
68214-500MG
68214-1G
68214-5G
68214-25G
77333-182EA
101.97
USD
77333-182
77343-014
77369-210
77381-296
(2,6-Difluoro-3-(methoxymethyl)phenyl)(methyl)sulfane ≥97%
(2,6-Difluoro-3-(methoxymethyl)phenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₉H₁₀F₂OS Storage Temperature: Ambient |
MDL Number:
MFCD34763244 CAS Number: 2807447-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...