1,3-Difluoro-2-iodo-4-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

68213-500MG 68213-1G 68213-5G 68213-25G
77333-180EA 108.15 USD
77333-180 77343-202 77369-208 77381-294
1,3-Difluoro-2-iodo-4-(methoxymethyl)benzene ≥95%
1,3-Difluoro-2-iodo-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34763243
CAS Number: 2901107-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR