2,6-Difluoro-3-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

68209-500MG 68209-1G 68209-5G 68209-25G
77333-178EA 143.17 USD
77333-178 77345-010 77369-206 77390-274
2,6-Difluoro-3-(methoxymethyl)benzoic acid ≥97%
2,6-Difluoro-3-(methoxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34763240
CAS Number: 1895589-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR