2-Bromo-6-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
68205-500MG
68205-1G
68205-5G
68205-25G
77333-172EA
121.54
USD
77333-172
77343-876
77369-200
77381-290
2-Bromo-6-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Bromo-6-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₉BrF₄O Storage Temperature: Ambient |
MDL Number:
MFCD34763237 CAS Number: 2901099-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...